Anti-mouse Adiponectin Receptor 1 Rabbit Polyclonal Antibody
Proveedor: Biosensis
BSENR-1329-100EA 345 EUR
BSENR-1329-100
Anti-mouse Adiponectin Receptor 1 Rabbit Polyclonal Antibody
Anticuerpos
Adiponectin Receptors 1 and 2  are membrane receptors for adiponectin, a hormone secreted by adipocytes which regulates energy homeostatis and insulin sensitivity.
Antibody recognizes Adiponectin Receptors 1 by western blot, other applications have not been tested. . A dilution of 1:2000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user. This antiserum is known to recognise both mouse and human Adiponectin Receptor 1.
Application Information:
Western Blotting. A dilution of 1:2000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: mouse Adiponectin Receptor 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype:
Reactivity: Mouse, Human