Anti-Parkin Rabbit Polyclonal Antibody

Proveedor: Biosensis
BSENR-114-100EA 334 EUR
BSENR-114-100
Anti-Parkin Rabbit Polyclonal Antibody
Anticuerpos
FUNCTION: Functions within a multiprotein E3 ubiquitin ligase complex, catalyzing the covalent attachment of ubiquitin moieties onto substrate proteins. These substrates include SYT11, CCNE1, GPR37, STUB1, a 22 kDa O-linked glycosylated isoform of SNCAIP and SEPT5. May play a more general role in the ubiquitin proteasomal pathway by participating in the removal and/or detoxification of abnormally folded or damaged protein. Loss of this ubiquitin ligase activity appears to be the mechanism underlying pathogenesis of PARK2. May protect neurons against alpha synuclein toxicity, proteasomal dysfunction, GPR37 accumulation, and kainate-induced excitotoxicity. May play a role in controlling neurotransmitter trafficking at the presynaptic terminal and in calcium-dependent exocytosis. Regulates cyclin E during neuronal apoptosis. May represent a tumor suppressor gene. SUBCELLULAR LOCATION: Cytoplasm. Co-localizes with STY11 in neutrites. Co-localizes with SNCAIP in brainstem Lewy bodies. TISSUE SPECIFICITY: Highly expressed in the brain including the substantia nigra. Expressed in heart, testis and skeletal muscle. Expression is down-regulated or absent in tumor biopsies, and absent in the brain of PARK2 patients. Overexpression protects dopamine neurons from kainate-mediated apoptosis.

This antiserum is known to be highly specific for Parkin shown by IHC and WB.
Application Information:
IHC, WB. A dilution of 1:1000 is recommended for immunohistochemistry and 1:2000 for western blot. Nice staining is achieved in neuronal and cytoplasmic granules sections treated with citrate buffer for antigen retrieval. Few inclusions are stained but these were not positively identified as Lewy bodies. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Parkin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR