Anti-ATG10 Rabbit Polyclonal Antibody

Proveedor: Biosensis
BSENR-145-100EA 334 EUR
BSENR-145-100
Anti-ATG10 Rabbit Polyclonal Antibody
Anticuerpos
ATG10 plays a role in autophagy. It is an E2-like enzyme involved in 2 ubiquitin-like modifications essential for autophagosome formation: ATG12-ATG5 conjugations and modificatioon of a soluble form of MAPLC3 1A, a homolog of yeast ATG8, to a membrane bound form.  It is also able to directly interact either with ATG5 or ATG7.

IHC, WB and ELISA confirmed the specificity for ATG10.
Application Information:
IHC, immunofluorescence. A dilution of 1:100 to 1:1000 dilution is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ATG10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal of Human ATG10
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR