Anti-Peripherin Rabbit Polyclonal Antibody

Proveedor: Biosensis
R-1401-50
BSENR-1401-50EA 302 EUR
BSENR-1401-50
Anti-Peripherin Rabbit Polyclonal Antibody
Anticuerpos
Peripherin is a class-III neuronal intermediate filament protein found in certain classes of neuron, most of which are located in the peripheral nervous system.

The specificity of this antibody has been confirmed by WB. This antibody detects ~57 kDa Peripherin protein and a smaller molecule derived from Peripherin at ~48 kDa.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). A dilution of 1:10,000 - 1:20,000 is recommended for WB. A dilution of 1:500-1,000 is recommended for IC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Peripherin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse, Cat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR