Neurofilaments contain three intermediate filament proteins: light (68 kDa), medium (160 kDa) and heavy (200 kDa). Neurofilament heavy (NF200 or NF-H) is phosphorylated and it is thought that this results in the formation of interfilament cross bridges that are important in the maintenance of axonal caliber.
The specificity of this antibody has been confirmed by WB. This antibody reacts with phosphorylated NF-H and is seen as a band of approx 200 kDa.
Application Information:
Western Blotting (WB), ELISA and Immunocytochemistry (IC). Suggested dilution for WB of 1:5,000-10,000. Suggested dilution for IC is 1:500-1,000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Neurofilament Heavy, phosphorylated
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Rat