Anti-dephosphorylated Catenin beta Sheep Polyclonal Antibody

Proveedor: Biosensis
BSENS-1233-100EA 387 EUR
BSENS-1233-100
Anti-dephosphorylated Catenin beta Sheep Polyclonal Antibody
Anticuerpos
Catenin beta is an adherens junction protein and has a role in the regulation of cell adhesion and in signal transduction through the Wnt pathway. At least 2 isoforms are produced by alternative splicing.

The specificity of this antibody has been confirmed by WB against the antigen.
Application Information:
Western Blotting (WB) and Immunocytochemistry. A concentration of 2.0 µg/ml is recommended for WB. Human Catenin beta (isoform 1) has a predicted length of 781 residues and MW of 86 kDa. It has been observed as a 92 kDa band in SDS-reducing gels. A concentration of 4.0 µg/ml is recommended for Immunocytochemistry. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: dephosphorylated Catenin beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acid region 28-46 of Human Catenin beta
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR