Anti-rh NT4 Sheep Polyclonal Antibody

Proveedor: Biosensis
BSENS-058-100EA 302 EUR
BSENS-058-100
Anti-rh NT4 Sheep Polyclonal Antibody
Anticuerpos
FUNCTION: Target-derived survival factor for peripheral sensory sympathetic neurons. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Highest levels in prostate, lower levels in thymus, placenta, and skeletal muscle. Expressed in embryonic and adult tissues. SIMILARITY: Belongs to the NGF-beta family.

Less than 1% cross-reactivity against NGF, recombinant human BDNF and 5% to NT3 has been shown by 1-site ELISA.
Application Information:
IHC, ELISA (1 site), Western Blot, dot blot, inhibition of biological activity in vitro/in vivo. Recommended to be used at a dilution of 1:500 to 1:2000 for immunohistochemistry, ELISA and Western blot. 1:10 to 1:50 for inhibition of biological activity in vitro. Use neat for in vivo studies at 5-10 µl/g body weight. Note that the concentration of NT4 is generally low in most tissues nevertheless, neonatal testes of rat can be used as a good positive control. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: rh NT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Sheep
Isotype: IgG
Reactivity: Rat, Human, Mouse, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR