Glutathione peroxidase 4 (GPx-4) is involved in protecting cells against membrane lipid peroxidation and cell death.
Human and rat mitochondrial glutathione peroxidase (GPx-4)
Application Information:
WB, IHC. Typical working dilution for IHC is 1:200 to 1:1,000 depending on tissue and detection method. For WB, a dilution range of 1:1,000 to 1:2,000 is recommended. GPx-4 exists as a tetramer and can reform into multimeric complexes even under reduced conditions. The reported molecular weight of the GPx-4 monomer is 22kDa but is reported to rapidly form oligomers, thus higher molecular weight bands greater than 22kDa are expected. Mitochondrial preparations are also recommended to enhance signal. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: glutathione peroxidase 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 78-93 of Human glutathione peroxidase 4
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat,