Anti-CHEK2 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29275EA 356 EUR
ABNOPAB29275
Anti-CHEK2 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CHEK2.

Recommended Dilutions: Immunofluorescence: 1:100~1:200; Western Blot: 1:500~1:1000

Type: Primary
Antigen: CHEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR