Anti-TNK2 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29267EA 356 EUR
ABNOPAB29267
Anti-TNK2 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human TNK2.

Recommended Dilutions: Immunofluorescence: 1:100~1:200; Immunohistochemistry: 1:50~1:100; Western Blot: 1:500~1:1000

Type: Primary
Antigen: TNK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR