Anti-FAM35A Mouse Monoclonal Antibody [clone: 2F9]

Proveedor: Abnova
ABNOH00054537-M08EA 394 EUR
ABNOH00054537-M08
Anti-FAM35A Mouse Monoclonal Antibody [clone: 2F9]
Anticuerpos
Mouse monoclonal antibody raised against a full-length recombinant FAM35A.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: FAM35A
Clonality: Monoclonal
Clone: 2F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR