Anti-HYKK Mouse Monoclonal Antibody [clone: 4F2]

Proveedor: Abnova
ABNOH00123688-M01EA 394 EUR
ABNOH00123688-M01
Anti-HYKK Mouse Monoclonal Antibody [clone: 4F2]
Anticuerpos
Mouse monoclonal antibody raised against a full-length recombinant HYKK.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: HYKK
Clonality: Monoclonal
Clone: 4F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR