Anti-RARRES2 Mouse Monoclonal Antibody [clone: 4B12]

Proveedor: Abnova
ABNOH00005919-M27JEA 558 EUR
ABNOH00005919-M27J
Anti-RARRES2 Mouse Monoclonal Antibody [clone: 4B12]
Anticuerpos
Mouse monoclonal antibody raised against a full-length recombinant RARRES2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: RARRES2
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR