Anti-RARRES2 Mouse Monoclonal Antibody [clone: 1G2]
Proveedor: Abnova
ABNOH00005919-M25JEA 558 EUR
ABNOH00005919-M25J
Anti-RARRES2 Mouse Monoclonal Antibody [clone: 1G2]
Anticuerpos
Mouse monoclonal antibody raised against a full-length recombinant RARRES2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: RARRES2
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now