Anti-BCAR1 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29258EA 419 EUR
ABNOPAB29258
Anti-BCAR1 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human BCAR1.

Recommended Dilutions: Immunohistochemistry: 1:50~1:100; Western Blot: 1:500~1:1000

Type: Primary
Antigen: BCAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR