Anti-CEBPA Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29256EA 419 EUR
ABNOPAB29256
Anti-CEBPA Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CEBPA.

Recommended Dilutions: Immunohistochemistry: 1:50~1:100; Western Blot: 1:500~1:1000

Type: Primary
Antigen: CEBPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR