Anti-ANKS3 Mouse Monoclonal Antibody [clone: 1B2]

Proveedor: Abnova
ABNOH00124401-M05EA 394 EUR
ABNOH00124401-M05
Anti-ANKS3 Mouse Monoclonal Antibody [clone: 1B2]
Anticuerpos
Mouse monoclonal antibody raised against a full-length recombinant ANKS3.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ANKS3
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR