Anti-SLC22A2 Mouse Monoclonal Antibody [clone: 1E3]

Proveedor: Abnova
ABNOH00006582-M02EA 394 EUR
ABNOH00006582-M02
Anti-SLC22A2 Mouse Monoclonal Antibody [clone: 1E3]
Anticuerpos
Mouse monoclonal antibody raised against a partial recombinant SLC22A2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SLC22A2
Clonality: Monoclonal
Clone: 1E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR