Anti-CFLAR Mouse Polyclonal Antibody
Proveedor: Abnova
ABNOH00008837-B02PEA 394 EUR
ABNOH00008837-B02P
Anti-CFLAR Mouse Polyclonal Antibody
Anticuerpos
Mouse polyclonal antibody raised against a full-length human CFLAR protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: CFLAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now