Anti-ATF2 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29235EA 356 EUR
ABNOPAB29235
Anti-ATF2 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human ATF2.

  • Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR