Anti-WWOX Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29231EA 356 EUR
ABNOPAB29231
Anti-WWOX Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human WWOX.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1000

Type: Primary
Antigen: WWOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR