Anti-EIF2AK3 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29217EA 419 EUR
ABNOPAB29217
Anti-EIF2AK3 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EIF2AK3.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: EIF2AK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR