Anti-IKBKG Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29299EA 356 EUR
ABNOPAB29299
Anti-IKBKG Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human IKBKG.

Recommended Dilutions: Immunohistochemistry: 1:50~1:100

Type: Primary
Antigen: IKBKG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR