Anti-IgG Chicken Polyclonal Antibody (Biotin)

Proveedor: Abnova
PAB29186
ABNOPAB29186EA 399 EUR
ABNOPAB29186
Anti-IgG Chicken Polyclonal Antibody (Biotin)
Anticuerpos
Chicken polyclonal antibody raised against native Human IgG F(ab')2 with Biotin conjugation.

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(ab')2
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR