Anti-IgY F(ab')2 Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: Abnova
ABNOPAB29159EA 370 EUR
ABNOPAB29159
Anti-IgY F(ab')2 Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Donkey anti-chicken IgY recognizes chicken IgY F(ab')2. The anti-F(ab')2-specific fraction was purified using antigen affinity chromatography.

Recommended Dilutions: Immunohistochemistry

Type: Secondary
Antigen: IgY F(ab')2
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(ab')2
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR