Anti-B4GALT2 Mouse Polyclonal Antibody
Proveedor: Abnova
ABNOH00008704-B01PEA 394 EUR
ABNOH00008704-B01P
Anti-B4GALT2 Mouse Polyclonal Antibody
Anticuerpos
Mouse polyclonal antibody raised against a full-length human B4GALT2 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: B4GALT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now