Anti-KDM5D Mouse Monoclonal Antibody [clone: 4C6]
Proveedor: Abnova
H00008284-M03
ABNOH00008284-M03EA 394 EUR
ABNOH00008284-M03
Anti-KDM5D Mouse Monoclonal Antibody [clone: 4C6]
Anticuerpos
Mouse monoclonal antibody raised against a partial recombinant JARID1D.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: KDM5D
Clonality: Monoclonal
Clone: 4C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now