Anti-GDF5 Mouse Monoclonal Antibody [clone: 1F4]

Proveedor: Abnova
ABNOH00008200-M06EA 394 EUR
ABNOH00008200-M06
Anti-GDF5 Mouse Monoclonal Antibody [clone: 1F4]
Anticuerpos
Mouse monoclonal antibody raised against a partial recombinant GDF5.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: GDF5
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR