Anti-ZNF28 Mouse Monoclonal Antibody [clone: 8H6]

Proveedor: Abnova
H00007576-M01
ABNOH00007576-M01EA 394 EUR
ABNOH00007576-M01
Anti-ZNF28 Mouse Monoclonal Antibody [clone: 8H6]
Anticuerpos
Mouse monoclonal antibody raised against a full-length recombinant ZNF28.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ZNF28
Clonality: Monoclonal
Clone: 8H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR