Anti-SCNN1A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Proveedor: Abnova
ABNOPAB28775EA 545 EUR
ABNOPAB28775
Anti-SCNN1A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Scnn1a.
Recommended Dilutions: Immunofluorescence: 1:400; Immunohistochemistry Western Blot: 1:1000
Type:
Antigen: SCNN1A
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now