Anti-SCNN1A Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28766EA 440 EUR
ABNOPAB28766
Anti-SCNN1A Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Rat Scnn1a.

Recommended Dilutions: Immunofluorescence: 1:400; Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SCNN1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR