Anti-SLC12A3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Proveedor: Abnova
ABNOPAB28758EA 545 EUR
ABNOPAB28758
Anti-SLC12A3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Human, Rat SLC12A3.

Recommended Dilutions: Immunofluorescence Western Blot: 1:1000

Type:
Antigen: SLC12A3
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR