Anti-SLC12A3 Rabbit Polyclonal Antibody (ATTO 488)
Proveedor: Abnova
ABNOPAB28750EA 545 EUR
ABNOPAB28750
Anti-SLC12A3 Rabbit Polyclonal Antibody (ATTO 488)
Anticuerpos
Rabbit polyclonal antibody raised against recombinant Human, Rat SLC12A3.
Recommended Dilutions: Immunofluorescence Western Blot: 1:1000
Type:
Antigen: SLC12A3
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now