Anti-ZNF558 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28745EA 545 EUR
ABNOPAB28745
Anti-ZNF558 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant ZNF558.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: ZNF558
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR