Anti-KCNA3 Mouse Monoclonal Antibody (ATTO 390) [clone: S23-27]

Proveedor: Abnova
ABNOMAB11657EA 545 EUR
ABNOMAB11657
Anti-KCNA3 Mouse Monoclonal Antibody (ATTO 390) [clone: S23-27]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of Kcna3.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: KCNA3
Clonality: Monoclonal
Clone: S23-27
Conjugation: ATTO 390
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR