Anti-PICK1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S20-8]

Proveedor: Abnova
MAB11636
ABNOMAB11636EA 545 EUR
ABNOMAB11636
Anti-PICK1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S20-8]
Anticuerpos
Mouse monoclonal antibody raised against partial synthetic protein of Pick1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: PICK1
Clonality: Monoclonal
Clone: S20-8
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR