Anti-TSPAN6 Mouse Monoclonal Antibody [clone: 1H1]
Proveedor: Abnova
ABNOH00007105-M06EA 394 EUR
ABNOH00007105-M06
Anti-TSPAN6 Mouse Monoclonal Antibody [clone: 1H1]
Anticuerpos
Mouse monoclonal antibody raised against a partial recombinant TSPAN6.
Type: Primary
Antigen: TSPAN6
Clonality: Monoclonal
Clone: 1H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now