Anti-SULT2B1 Mouse Polyclonal Antibody
Proveedor: Abnova
ABNOH00006820-B01PEA 394 EUR
ABNOH00006820-B01P
Anti-SULT2B1 Mouse Polyclonal Antibody
Anticuerpos
Mouse polyclonal antibody raised against a full-length human SULT2B1 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: SULT2B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now