Anti-SCD Mouse Monoclonal Antibody [clone: 1B7]

Proveedor: Abnova
ABNOH00006319-M02AEA 394 EUR
ABNOH00006319-M02A
Anti-SCD Mouse Monoclonal Antibody [clone: 1B7]
Anticuerpos
Mouse monoclonal antibody raised against a partial recombinant SCD.

Type: Primary
Antigen: SCD
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR