Anti-MAPK12 Mouse Monoclonal Antibody [clone: 2C1]

Proveedor: Abnova
ABNOH00006300-M04AEA 394 EUR
ABNOH00006300-M04A
Anti-MAPK12 Mouse Monoclonal Antibody [clone: 2C1]
Anticuerpos
Mouse monoclonal antibody raised against a partial recombinant MAPK12.

Type: Primary
Antigen: MAPK12
Clonality: Monoclonal
Clone: 2C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR