Anti-ABLIM3 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28724EA 545 EUR
ABNOPAB28724
Anti-ABLIM3 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant ABLIM3.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20; Western Blot: 1:100-1:500

Type: Primary
Antigen: ABLIM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR