Anti-PICK1 Mouse Monoclonal Antibody (ATTO 390) [clone: S20-8]
Proveedor: Abnova
ABNOMAB11621EA 545 EUR
ABNOMAB11621
Anti-PICK1 Mouse Monoclonal Antibody (ATTO 390) [clone: S20-8]
Anticuerpos
Mouse monoclonal antibody raised against partial synthetic protein of Pick1.
Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: PICK1
Clonality: Monoclonal
Clone: S20-8
Conjugation: ATTO 390
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now