Anti-OLFM1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S96-7]

Proveedor: Abnova
MAB11617
ABNOMAB11617EA 545 EUR
ABNOMAB11617
Anti-OLFM1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S96-7]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of OLFM1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: OLFM1
Clonality: Monoclonal
Clone: S96-7
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR