Anti-OLFM1 Mouse Monoclonal Antibody (ATTO 488) [clone: S96-7]
Proveedor: Abnova
ABNOMAB11604EA 545 EUR
ABNOMAB11604
Anti-OLFM1 Mouse Monoclonal Antibody (ATTO 488) [clone: S96-7]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of OLFM1.
Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: OLFM1
Clonality: Monoclonal
Clone: S96-7
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now