Anti-RAB3A Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28701EA 545 EUR
ABNOPAB28701
Anti-RAB3A Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant RAB3A.

Recommended Dilutions: Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: RAB3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR