Anti-BGN Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28699EA 545 EUR
ABNOPAB28699
Anti-BGN Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant BGN.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Western Blot: 1:250-1:500

Type: Primary
Antigen: BGN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR