Anti-FBXO10 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28695EA 545 EUR
ABNOPAB28695
Anti-FBXO10 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant FBXO10.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: FBXO10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR