Anti-RPS6KA6 Rabbit Polyclonal Antibody

Proveedor: Abnova
PAB28694
ABNOPAB28694EA 518 EUR
ABNOPAB28694
Anti-RPS6KA6 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant RPS6KA6.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: RPS6KA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR