Anti-UQCRC1 Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28688EA 518 EUR
ABNOPAB28688
Anti-UQCRC1 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant UQCRC1.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Immunofluorescence: 1-4 ?g/ml; Western Blot: 1:100-1:500

Type: Primary
Antigen: UQCRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR