Anti-AURKA Rabbit Polyclonal Antibody

Proveedor: Abnova
ABNOPAB28676EA 518 EUR
ABNOPAB28676
Anti-AURKA Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal antibody raised against recombinant AURKA.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:100-1:250

Type: Primary
Antigen: AURKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR